Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027444.3 | NP_081720.2 | 5 | 617 | |
GenBank mRNA | AF276950.1 | - | 5 | 502 | |
AF276951.1 | - | 5 | 589 | ||
AF276952.1 | - | 2 | 345 | ||
AF454943.1 | - | 5 | 610 | ||
AF454944.1 | - | 3 | 482 | ||
AK017487.1 | - | 5 | 610 | ||
AK029532.1 | - | 3 | 484 | ||
AK029747.1 | - | 6 | 731 | ||
AK049516.1 | - | 5 | 603 | ||
AK050488.1 | - | 5 | 556 | ||
AK157813.1 | - | 5 | 596 | ||
AK167724.1 | - | 5 | 748 | ||
BC019456.1 | - | 5 | 692 | ||
BC024449.1 | - | 3 | 465 | ||
BC038874.1 | - | 1 | 156 | ||
BC057869.1 | - | 5 | 585 |