Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027909.2 | NP_082185.2 | 13 | 2244 | |
GenBank mRNA | AK004920.1 | - | 13 | 2249 | |
AK039220.1 | - | 13 | 2215 | ||
AK122237.1 | - | 8 | 3819 | ||
AK153804.1 | - | 13 | 2216 | ||
BC035271.1 | - | 10 | 2250 | ||
BC049875.1 | - | 9 | 1171 | ||
BC056612.1 | - | 13 | 1815 | 4952570 | |
BC060156.1 | - | 13 | 2108 | 6810220 |