Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001285839.1 | NP_001272768.1 | 4 | 987 | |
GenBank mRNA | AK011265.1 | - | 4 | 935 | |
AK045669.1 | - | 4 | 949 | ||
AK137332.1 | - | 4 | 938 | ||
BC030671.1 | - | 4 | 952 | ||
BC038910.1 | - | 2 | 313 | ||
BC051211.1 | - | 1 | 363 | ||
BC058172.1 | - | 4 | 950 | 5012054 |