Gene Transcripts

Gene Symbol
Atxn7l1
Entrez Gene ID
380753
Gene Name
ataxin 7-like 1
Gene Aliases
2810423G08Rik, Atxn7l4
Chromosome Location
Chr. 12: 33147639 - 33376408 on Build GRCm38
UniGene ID
Mm.352037
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028139.5 NP_082415.2 1, 2 231
GenBank mRNA AK013145.1 - 1, 2 138
BB843098.1 - 1, 2 154
BY203850.1 - 1, 2 103

View Full Product Details