Gene Transcripts

Gene Symbol
Smchd1
Entrez Gene ID
74355
Gene Name
SMC hinge domain containing 1
Gene Aliases
4931400A14Rik, AW554188, MommeD1, mKIAA0650
Chromosome Location
Chr. 17: 71344489 - 71475343 on Build GRCm38
UniGene ID
Mm.194450
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028887.3 NP_083163.3 23, 24 3086
GenBank mRNA AK129181.1 - 1, 2 83
BC062946.1 - 9, 10 931 30357469
BC076627.1 - 16, 17 2049

View Full Product Details