Gene Transcripts

Gene Symbol
Ropn1
Entrez Gene ID
76378
Gene Name
ropporin, rhophilin associated protein 1
Gene Aliases
1700008N21Rik, ODF6, RHPNAP1, Rppn, ropporin
Chromosome Location
Chr. 16: 34649842 - 34678610 on Build GRCm38
UniGene ID
Mm.23515
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030744.2 NP_109669.1 5 569
GenBank mRNA AF178531.1 - 4 492
AK160142.1 - 3 311
BC049544.1 - 5 569 6740976

View Full Product Details