Gene Transcripts
- Gene Symbol
- Arsk
- Entrez Gene ID
- 77041
- Gene Name
- arylsulfatase K
- Gene Aliases
- 2810429K17Rik, 4833414G15Rik
- Chromosome Location
- Chr. 13: 76060422 - 76098667 on Build GRCm38
- UniGene ID
- Mm.196399
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029847.4 |
NP_084123.2
|
3, 4 |
569 |
|
GenBank mRNA |
AK013194.2 |
- |
3, 4 |
521 |
|
AK019515.1 |
- |
3, 4 |
535 |
|
AK031147.1 |
- |
3, 4 |
557 |
|
AK032812.1 |
- |
3, 4 |
535 |
|
AK033968.1 |
- |
3, 4 |
570 |
|
AK035464.1 |
- |
3, 4 |
490 |
|
AK039765.1 |
- |
3, 4 |
565 |
|
AK046378.1 |
- |
3, 4 |
570 |
|
AK083188.1 |
- |
3, 4 |
523 |
|
AK084090.1 |
- |
3, 4 |
555 |
|
AK086667.1 |
- |
3, 4 |
528 |
|
AK144817.1 |
- |
3, 4 |
537 |
|
AK165970.1 |
- |
3, 4 |
537 |
|
BC046790.1 |
- |
3, 4 |
543 |
5717223 |
BC058351.1 |
- |
3, 4 |
543 |
5717223 |
View Full Product Details