Gene Transcripts
- Gene Symbol
- Spsb3
- Entrez Gene ID
- 79043
- Gene Name
- splA/ryanodine receptor domain and SOCS box containing 3
- Gene Aliases
- 2310012N15Rik, 3300001M01Rik, SSB3, Tce1, mKIAA4204
- Chromosome Location
- Chr. 17: 24886349 - 24892147 on Build GRCm38
- UniGene ID
- Mm.274794
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF403038.1 |
- |
7 |
855 |
|
AK009325.1 |
- |
7 |
855 |
|
View Full Product Details