Gene Transcripts

Gene Symbol
Bcl9l
Entrez Gene ID
80288
Gene Name
B cell CLL/lymphoma 9-like
Gene Aliases
B9L, BC003321, BCL9-2, DLNB11
Chromosome Location
Chr. 9: 44483113 - 44511901 on Build GRCm38
UniGene ID
Mm.370270
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030256.2 NP_084532.2 3 480
GenBank mRNA AB128033.1 - 2 266
AK155681.1 - 4 440
AY296058.1 - 3 480
BC082304.1 - 2 614 6856460

View Full Product Details