Gene Transcripts

Gene Symbol
Trim6
Entrez Gene ID
94088
Gene Name
tripartite motif-containing 6
Gene Aliases
C430046K18Rik, D7Ertd684e
Chromosome Location
Chr. 7: 104218783 - 104235152 on Build GRCm38
UniGene ID
Mm.274843
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013616.2 NP_001013637.1 4, 5 898
GenBank mRNA AK033396.1 - 4, 5 878
AK049238.1 - 4, 5 878
BC138965.1 - 4, 5 829 8861987

View Full Product Details