Gene Transcripts

Gene Symbol
Ankrd10
Entrez Gene ID
102334
Gene Name
ankyrin repeat domain 10
Gene Aliases
4833425P12Rik, AW549277
Chromosome Location
Chr. 8: 11611581 - 11636136 on Build GRCm38
UniGene ID
Mm.12459
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167967.2 NP_001161439.1 4 759
NM_001281974.1 NP_001268903.1 5 871
NM_001281975.1 NP_001268904.1 5 921
NM_133971.3 NP_598732.2 4 759
GenBank mRNA AK014768.1 - 5 750
AK031808.1 - 4 757
AK033623.1 - 4 757
AK038156.1 - 4 757
AK039879.1 - 2 204
AK042500.1 - 4 757
AK050983.1 - 4 760
AK081754.1 - 5 919
AK082875.1 - 4 756
AK084456.1 - 5 869
AK086012.1 - 2 1114
AK088371.1 - 4 709
BC002198.1 - 4 691 3489133
BC033266.1 - 1 54

View Full Product Details