Gene Transcripts

Gene Symbol
Ano10
Entrez Gene ID
102566
Gene Name
anoctamin 10
Gene Aliases
AI604832, Tmem16k
Chromosome Location
Chr. 9: 122175874 - 122294443 on Build GRCm38
UniGene ID
Mm.87676
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271873.1 NP_001258802.1 2 189
NM_133979.3 NP_598740.1 2 189
GenBank mRNA AK043011.1 - 2 152
AK046091.1 - 2 141
AK079622.1 - 2 140
AK080947.1 - 2 141
AK138414.1 - 2 175
BC099688.1 - 2 112 6313494

View Full Product Details