Gene Transcripts

Gene Symbol
Tbc1d10a
Entrez Gene ID
103724
Gene Name
TBC1 domain family, member 10a
Gene Aliases
AI447804, EPI64, Tbc1d10, mFLJ00288
Chromosome Location
Chr. 11: 4186833 - 4215507 on Build GRCm38
UniGene ID
Mm.28140
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134023.1 NP_598784.1 3 443
GenBank mRNA AK140872.1 - 3 455
AK153677.1 - 425
AK197604.1 - 1 35
AK220243.1 - 454
BC018300.1 - 3 443 4193046
BC037230.1 - 3 426 2650923

View Full Product Details