Gene Transcripts

Gene Symbol
Rcbtb2
Entrez Gene ID
105670
Gene Name
regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2
Gene Aliases
2610028E02Rik, 2810420M18Rik, AI448345, AW240694, Chc1l, RC/BTB2
Chromosome Location
Chr. 14: 73138773 - 73184055 on Build GRCm38
UniGene ID
Mm.280068
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170694.1 NP_001164165.1 8 844
NM_134083.4 NP_598844.3 8 839
GenBank mRNA AK013540.1 - 948
AK031242.1 - 8 805
AK031248.1 - 8 810
AK147784.1 - 9 845
AK160881.1 - 9 1031
BC003224.1 - 6 637 3482729
JN086338.1 - 7 760
JN086339.1 - 8 998

View Full Product Details