Gene Transcripts

Gene Symbol
Vmac
Entrez Gene ID
106639
Gene Name
vimentin-type intermediate filament associated coiled-coil protein
Gene Aliases
AI662250
Chromosome Location
Chr. 17: 56713932 - 56717699 on Build GRCm38
UniGene ID
Mm.37290
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166474.1 NP_001159946.1 2 543
NM_178926.3 NP_849257.1 2 729
GenBank mRNA AK078549.1 - 2 660
AK145854.1 - 2 582
AK170944.1 - 2 675
BC026472.1 - 2 729 4487505
BC039944.1 - 2 507
BC058424.1 - 2 524

View Full Product Details