Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024191.2 | NP_077153.1 | 3 | 378 | |
NM_024269.4 | NP_077231.1 | 3 | 216 | ||
GenBank mRNA | AK006418.1 | - | 3 | 203 | |
AK006646.1 | - | 3 | 202 | ||
AK018229.1 | - | 3 | 188 | ||
AK149620.1 | - | 3 | 216 | ||
AK150513.1 | - | 3 | 380 | ||
AK155242.1 | - | 3 | 329 | ||
AK161325.1 | - | 3 | 197 | ||
BC002131.1 | - | 3 | 223 | 3592643 | |
BC024708.1 | - | 3 | 327 | 3987328 | |
CT010168.1 | - | 3 | 114 |