Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028882.4 | NP_083158.3 | 4 | 888 | |
GenBank mRNA | AK028900.1 | - | 4 | 670 | |
AK031704.1 | - | 4 | 500 | ||
AK051165.1 | - | 4 | 549 | ||
AK052671.1 | - | 4 | 889 | ||
AK134412.1 | - | 4 | 524 | ||
AK153839.1 | - | 4 | 549 | ||
BC138131.1 | - | 4 | 431 | 8861150 |