Gene Transcripts

Gene Symbol
Slc9a4
Entrez Gene ID
110895
Gene Name
solute carrier family 9 (sodium/hydrogen exchanger), member 4
Gene Aliases
AW990558, D730009J23Rik, NHE4
Chromosome Location
Chr. 1: 40579715 - 40630731 on Build GRCm38
UniGene ID
Mm.108602
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177084.3 NP_796058.1 1 354
GenBank mRNA AK085681.1 - 1 380
AK135750.1 - 1 381
AK141534.1 - 1 355
BC120543.1 - 1 272 40129466

View Full Product Details