Gene Transcripts

Gene Symbol
A630095E13Rik
Entrez Gene ID
235973
Gene Name
RIKEN cDNA A630095E13 gene
Gene Aliases
Gm191, Sslp-1, Sslp1
Chromosome Location
Chr. 9: 36635754 - 36638602 on Build GRCm38
UniGene ID
Mm.118804
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033325.2 NP_001028497.1 2 212
GenBank mRNA AK042477.1 - 301
AK144443.1 - 2 213
BB873517.1 - 2 213
CJ069767.1 - 2 213

View Full Product Details