Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153469.4 | NP_703199.1 | 3 | 924 | |
XM_006235518.2 | XP_006235580.1 | 3 | 2165 | ||
XM_006235520.2 | XP_006235582.1 | 4 | 1016 | ||
XM_006235521.1 | XP_006235583.1 | 4 | 1108 | ||
XM_006235522.1 | XP_006235584.1 | 3 | 1039 | ||
XM_008762469.1 | XP_008760691.1 | 3 | 2386 | ||
XM_008762470.1 | XP_008760692.1 | 3 | 955 | ||
GenBank mRNA | BC088108.1 | - | 3 | 924 | |
FQ218619.1 | - | 3 | 1020 |