Gene Transcripts

Gene Symbol
EDN2
Entrez Gene ID
1907
Gene Name
endothelin 2
Gene Aliases
ET-2, ET2, PPET2
Chromosome Location
Chr. 1: 41478775 - 41484699 on Build GRCh38
UniGene ID
Hs.1407
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302269.1 NP_001289198.1 4 1130
NM_001956.4 NP_001947.1 5 1229
NR_126098.1 5 1138
XM_017000512.1 XP_016856001.1 3 1022
GenBank mRNA BC034393.1 - 5 1172
M65199.1 - 5 1217
X55177.1 - 5 1205

View Full Product Details