Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024496.3 | NP_078772.1 | 1 | 3541 | |
GenBank mRNA | AB058768.1 | - | 2 | 3036 | |
AF063597.1 | - | 1 | 671 | ||
AL832320.1 | - | 1 | 1643 | ||
BC108292.1 | - | 1 | 3538 | ||
BC152431.1 | - | 2 | 3036 |