Gene Transcripts

Gene Symbol
ULK2
Entrez Gene ID
9706
Gene Name
unc-51 like autophagy activating kinase 2
Gene Aliases
ATG1B, Unc51.2
Chromosome Location
Chr. 17: 19770828 - 19867926 on Build GRCh38
UniGene ID
Hs.168762
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142610.1 NP_001136082.1 8 1113
NM_014683.3 NP_055498.3 8 1113
XM_011524087.2 XP_011522389.1 4 410
XM_017025424.1 XP_016880913.1 8 1001
XM_017025425.1 XP_016880914.1 8 1001
XM_017025426.1 XP_016880915.1 8 1001
XM_017025427.1 XP_016880916.1 7 514
XM_017025428.1 XP_016880917.1 7 514
GenBank mRNA AB014523.1 - 8 1104
AK098316.1 - 7 738
AK314345.1 - 8 638
BC034988.1 - 9 678 5268025

View Full Product Details