Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001202502.1 | NP_001189431.1 | 3 | 202 | |
NM_003634.3 | NP_003625.2 | 3 | 503 | ||
GenBank mRNA | AJ001258.1 | - | 3 | 502 | |
AK098367.1 | - | 3 | 257 | ||
AK298816.1 | - | 3 | 204 | ||
AK314407.1 | - | 3 | 274 | ||
BC002371.2 | - | 3 | 253 | 2822369 | |
BC006473.1 | - | 3 | 253 | 2822369 | |
BC018869.2 | - | 2 | 145 | ||
DA133702.1 | - | 3 | 321 |