Gene Transcripts

Gene Symbol
PEX11B
Entrez Gene ID
8799
Gene Name
peroxisomal biogenesis factor 11 beta
Gene Aliases
PEX11-BETA, PEX14B
Chromosome Location
Chr. 1: 145911348 - 145918924 on Build GRCh38
UniGene ID
Hs.714608
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184795.1 NP_001171724.1 4 1609
NM_003846.2 NP_003837.1 4 1803
NR_073491.1 4 1799
NR_073492.1 4 1793
NR_073493.2 4 2015
GenBank mRNA AB018080.1 - 4 1585
AF093670.1 - 4 1583
AK023991.1 - 4 1567
AK094173.1 - 1 2403
BC011963.1 - 4 1583 3866690

View Full Product Details