Gene Transcripts

Gene Symbol
Ppan
Entrez Gene ID
235036
Gene Name
peter pan homolog (Drosophila)
Gene Aliases
A230087P06Rik, SSF1
Chromosome Location
Chr. 9: 20888175 - 20892179 on Build GRCm38
UniGene ID
Mm.2930
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145610.2 NP_663585.2 11 1636
GenBank mRNA AK039027.1 - 11 1541
AK085695.1 - 1 3880
AK171672.1 - 7 1302
AK172615.1 - 7 1302
BB749945.1 - 2 401
BC014688.1 - 11 1554 4489442
BC059277.1 - 9 1800

View Full Product Details