Gene Transcripts

Gene Symbol
Pex11a
Entrez Gene ID
18631
Gene Name
peroxisomal biogenesis factor 11 alpha
Gene Aliases
PEX11alpha
Chromosome Location
Chr. 7: 79735957 - 79743126 on Build GRCm38
UniGene ID
Mm.20615
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011068.1 NP_035198.1 2, 3 226
GenBank mRNA AF093669.1 - 2, 3 226
AK078954.1 - 1, 2 59
AK149387.1 - 2, 3 224
BC023439.1 - 2, 3 199 5251140

View Full Product Details