Gene Transcripts
- Gene Symbol
- Slc25a22
- Entrez Gene ID
- 68267
- Gene Name
- solute carrier family 25 (mitochondrial carrier, glutamate), member 22
- Gene Aliases
- 1300006L01Rik, AI060884, Gc1
- Chromosome Location
- Chr. 7: 141429749 - 141437887 on Build GRCm38
- UniGene ID
- Mm.33729
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001177576.1 |
NP_001171047.1
|
8, 9 |
958 |
|
NM_026646.3 |
NP_080922.1
|
8, 9 |
1200 |
|
GenBank mRNA |
AK010193.1 |
- |
8, 9 |
946 |
|
AK018760.1 |
- |
8, 9 |
983 |
|
AK046874.1 |
- |
8, 9 |
945 |
|
AK048389.1 |
- |
8, 9 |
929 |
|
AK049313.1 |
- |
8, 9 |
929 |
|
AK155276.1 |
- |
8, 9 |
952 |
|
AK162400.1 |
- |
8, 9 |
911 |
|
BC037949.1 |
- |
8, 9 |
957 |
5367108 |
BC050887.1 |
- |
7, 8 |
1676 |
|
View Full Product Details