Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152804.2 | NP_690017.2 | 7 | 1010 | |
GenBank mRNA | AK133962.1 | - | 7 | 1009 | |
AK164365.1 | - | 7 | 1009 | ||
BC034513.1 | - | 7 | 992 | 3156743 | |
BC035451.1 | - | 7 | 992 | ||
M96163.1 | - | 7 | 976 |