Gene Transcripts

Gene Symbol
Btnl10
Entrez Gene ID
192194
Gene Name
butyrophilin-like 10
Gene Aliases
BUTR-1, Butr1
Chromosome Location
Chr. 11: 58917878 - 58926965 on Build GRCm38
UniGene ID
Mm.22540
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138678.2 NP_619619.2 5 861
GenBank mRNA AF269232.1 - 5 861
AK029784.1 - 7 2544

View Full Product Details