Gene Transcripts

Gene Symbol
Tas2r107
Entrez Gene ID
387342
Gene Name
taste receptor, type 2, member 107
Gene Aliases
T2R07, T2R4, Tas2r7, mGR06, mt2r43
Chromosome Location
Chr. 6: 131659158 - 131660084 on Build GRCm38
UniGene ID
Mm.349283
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199154.1 NP_954605.1 1 411
GenBank mRNA AF412301.1 - 1 246
BC141027.1 - 1 476 9055639

View Full Product Details