Gene Transcripts

Gene Symbol
Reep2
Entrez Gene ID
225362
Gene Name
receptor accessory protein 2
Gene Aliases
Chromosome Location
Chr. 18: 34840589 - 34847463 on Build GRCm38
UniGene ID
Mm.293588
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204914.1 NP_001191843.1 8 1340
NM_144865.2 NP_659114.2 8 1346
GenBank mRNA AK147333.1 - 2 6136
AK158694.1 - 3 920
BC020184.1 - 8 1277 4505679

View Full Product Details