Gene Transcripts

Gene Symbol
VAT1
Entrez Gene ID
10493
Gene Name
vesicle amine transport 1
Gene Aliases
VATI
Chromosome Location
Chr. 17: 43014605 - 43022442 on Build GRCh38
UniGene ID
Hs.514199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006373.3 NP_006364.2 2 605
GenBank mRNA AK097072.1 - 3 432
AK290460.1 - 2 581
AK298536.1 - 2 315
AK315838.1 - 2 169
BC001913.1 - 2 474
BC008725.2 - 2 529 3162543
BC014279.2 - 2 511 3834206
BC015041.2 - 2 543 3919901
BC090858.1 - 2 543 6528247
BT007369.1 - 2 485
U18009.1 - 2 243
U25779.1 - 2 209

View Full Product Details