Gene Transcripts

Gene Symbol
Ttc29
Entrez Gene ID
73301
Gene Name
tetratricopeptide repeat domain 29
Gene Aliases
1700031F13Rik
Chromosome Location
Chr. 8: 78213302 - 78394326 on Build GRCm38
UniGene ID
Mm.61243
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183096.3 NP_898919.3 4, 5 491
GenBank mRNA AK006580.1 - 4, 5 434
BC048386.1 - 4, 5 491 6310247

View Full Product Details