Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022025.4 | NP_071308.2 | 5 | 978 | |
GenBank mRNA | AF276872.2 | - | 4 | 469 | |
AJ401467.1 | - | 4 | 469 | ||
AK034415.1 | - | 5 | 862 | ||
AK053063.1 | - | 5 | 980 | ||
BC065089.1 | - | 4 | 495 | 6851099 |