Gene Transcripts

Gene Symbol
Plac1
Entrez Gene ID
56096
Gene Name
placental specific protein 1
Gene Aliases
DXWsu72e, Epcs26, Oosp2l
Chromosome Location
Chr. X: 53069993 - 53240122 on Build GRCm38
UniGene ID
Mm.274842
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019538.4 NP_062411.1 3 314
GenBank mRNA AF234653.1 - 3 315
AF250838.1 - 3 311
AK005501.1 - 3 315
AK145876.1 - 3 280
BC051666.1 - 3 300 30017089

View Full Product Details