Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098783.2 | NP_001092253.1 | 6 | 892 | |
NM_007042.4 | NP_008973.1 | 6 | 658 | ||
NR_049755.1 | 5 | 794 | |||
NR_049756.1 | 6 | 658 | |||
NR_049757.1 | 5 | 560 | |||
NR_049758.1 | 4 | 475 | |||
XM_011533313.2 | XP_011531615.1 | 6 | 692 | ||
XM_017005641.1 | XP_016861130.1 | 6 | 655 | ||
GenBank mRNA | AF001175.1 | - | 6 | 634 | |
AK311396.1 | - | 5 | 519 | ||
BC002441.1 | - | 6 | 606 | 3347003 | |
BC007342.1 | - | 6 | 606 | 3616325 | |
BC012017.1 | - | 6 | 619 | 4708724 |