Gene Transcripts

Gene Symbol
MKRN2
Entrez Gene ID
23609
Gene Name
makorin ring finger protein 2
Gene Aliases
HSPC070, RNF62
Chromosome Location
Chr. 3: 12557014 - 12583713 on Build GRCh38
UniGene ID
Hs.591666
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014160.4 NP_054879.3 2 271
GenBank mRNA AF302084.1 - 2 185
AK091318.1 - 2 216
BC015715.1 - 2 213 3916199
DR157266.1 - 2 271

View Full Product Details