Gene Transcripts

Gene Symbol
SMPX
Entrez Gene ID
23676
Gene Name
small muscle protein, X-linked
Gene Aliases
DFN6, DFNX4
Chromosome Location
Chr. X: 21705972 - 21758160 on Build GRCh38
UniGene ID
Hs.734084
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014332.2 NP_055147.1 5 756
NR_045617.1 6 858
GenBank mRNA AF129505.1 - 5 708
AJ250584.1 - 5 708
BC005948.1 - 5 637 4246501

View Full Product Details