Gene Transcripts

Gene Symbol
Nup35
Entrez Gene ID
295692
Gene Name
nucleoporin 35
Gene Aliases
Chromosome Location
Chr. 3: 67866830 - 67896828 on Build Rnor_6.0
UniGene ID
Rn.203627
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004229.1 NP_001004229.1 7 710
XM_006234430.2 XP_006234492.1 7 717
GenBank mRNA BC078914.1 - 7 710

View Full Product Details