Gene Transcripts

Gene Symbol
THEM5
Entrez Gene ID
284486
Gene Name
thioesterase superfamily member 5
Gene Aliases
ACOT15
Chromosome Location
Chr. 1: 151847101 - 151853697 on Build GRCh38
UniGene ID
Hs.132648
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182578.3 NP_872384.1 5 770
GenBank mRNA AK095283.1 - 5 770
BC101610.1 - 5 676 8069116
BC112239.1 - 5 676 8327707

View Full Product Details