Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001324371.1 | NP_001311300.1 | 3 | 404 | |
NM_001324372.1 | NP_001311301.1 | 3 | 408 | ||
NM_001324373.1 | NP_001311302.1 | 3 | 408 | ||
NM_017888.2 | NP_060358.2 | 3 | 408 | ||
XM_006721056.3 | XP_006721119.1 | 3 | 430 | ||
GenBank mRNA | AK000588.1 | - | 3 | 405 | |
BC013753.1 | - | 3 | 396 | ||
BC016703.1 | - | 3 | 412 | ||
BC068516.1 | - | 3 | 408 | 5261244 |