Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029320.3 | NP_083596.2 | 8, 9 | 1826 | |
GenBank mRNA | AK030385.1 | - | 8, 9 | 1307 | |
AK149007.1 | - | 3, 4 | 379 | ||
BC129811.1 | - | 8, 9 | 1303 | 40062382 | |
BC129812.1 | - | 8, 9 | 1303 | 40062383 |