Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029932.3 | NP_084208.1 | 4 | 509 | |
GenBank mRNA | AK020683.1 | - | 4 | 510 | |
AK041376.1 | - | 3 | 499 | ||
AK156479.1 | - | 4 | 773 | ||
AK156781.1 | - | 4 | 502 | ||
BC150778.1 | - | 4 | 492 | 9087689 |