Gene Transcripts

Gene Symbol
SFTA3
Entrez Gene ID
253970
Gene Name
surfactant associated 3
Gene Aliases
NANCI, SFTPH, SP-H
Chromosome Location
Chr. 14: 36473288 - 36513785 on Build GRCh38
UniGene ID
Hs.509165
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101341.1 NP_001094811.1 1, 2 679
GenBank mRNA BC042093.1 - 1, 2 679
BC129834.1 - 1, 2 425
KF856952.1 - 1, 2 774

View Full Product Details