Gene Transcripts

Gene Symbol
XRRA1
Entrez Gene ID
143570
Gene Name
X-ray radiation resistance associated 1
Gene Aliases
Chromosome Location
Chr. 11: 74840910 - 74949187 on Build GRCh38
UniGene ID
Hs.370145
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270380.1 NP_001257309.1 15 2870
NM_001270381.1 NP_001257310.1 16 2969
NM_182969.2 NP_892014.1 19 3237
XM_005273765.2 XP_005273822.1 19 3261
XM_005273770.2 XP_005273827.1 17 3041
XM_011544755.1 XP_011543057.1 19 3216
XM_011544756.1 XP_011543058.1 18 3135
XM_011544757.1 XP_011543059.1 18 3090
XM_011544758.1 XP_011543060.1 17 3090
XM_011544759.1 XP_011543061.1 17 3066
XM_011544761.1 XP_011543063.1 16 3018
XM_011544762.1 XP_011543064.1 16 3000
XM_011544763.1 XP_011543065.1 18 3155
XM_011544764.1 XP_011543066.1 16 2925
XM_011544765.1 XP_011543067.1 17 2975
XM_011544766.2 XP_011543068.1 19 3280
XM_011544767.1 XP_011543069.1 18 3182
XM_011544768.1 XP_011543070.1 17 3029
XM_017017221.1 XP_016872710.1 17 3051
XM_017017222.1 XP_016872711.1 17 3029
XM_017017223.1 XP_016872712.1 16 2984
XM_017017224.1 XP_016872713.1 15 2894
XM_017017225.1 XP_016872714.1 15 2819
XM_017017226.1 XP_016872715.1 18 3156
XM_017017227.1 XP_016872716.1 18 3132
XM_017017228.1 XP_016872717.1 16 2935
XM_017017229.1 XP_016872718.1 17 2985
XM_017017230.1 XP_016872719.1 16 2809
XM_017017231.1 XP_016872720.1 15 2895
XM_017017232.1 XP_016872721.1 13 2599
XM_017017233.1 XP_016872722.1 16 2989
XM_017017234.1 XP_016872723.1 15 2874
GenBank mRNA AK074152.1 - 14 2567
BC037294.2 - 16 2902
BC040541.1 - 16 2902
CR749605.1 - 14 3341

View Full Product Details