Gene Transcripts

Gene Symbol
C7orf71
Entrez Gene ID
285941
Gene Name
chromosome 7 open reading frame 71
Gene Aliases
Chromosome Location
Chr. 7: 26635582 - 26647272 on Build GRCh38
UniGene ID
Hs.413394
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145531.2 NP_001139003.1 4 1607
NM_001302625.1 NP_001289554.1 3 947
XM_017012010.1 XP_016867499.1 4 4097
XM_017012011.1 XP_016867500.1 5 3983
XM_017012012.1 XP_016867501.1 4 3709
XM_017012013.1 XP_016867502.1 5 3625
XM_017012014.1 XP_016867503.1 4 3705
GenBank mRNA AK093321.1 - 4 1607
CA390440.1 - 1 74

View Full Product Details