Gene Transcripts

Gene Symbol
PRCD
Entrez Gene ID
768206
Gene Name
progressive rod-cone degeneration
Gene Aliases
RP36
Chromosome Location
Chr. 17: 76527586 - 76555339 on Build GRCh38
UniGene ID
Hs.634380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077620.2 NP_001071088.1 4, 5 414
NR_033357.1 4, 5 485
XM_017025013.1 XP_016880502.1 4, 5 2432
XM_017025014.1 XP_016880503.1 4, 5 2432
GenBank mRNA AK125617.1 - 4, 5 485
BC043413.1 - 4, 5 282
BC146898.1 - 4, 5 355 9056826
BC146904.1 - 4, 5 355 9056832

View Full Product Details