Gene Transcripts

Gene Symbol
NBPF15
Entrez Gene ID
284565
Gene Name
neuroblastoma breakpoint family member 15
Gene Aliases
AB14, AG3, NBPF16
Chromosome Location
Chr. 1: 144421386 - 144459464 on Build GRCh38
UniGene ID
Hs.523572
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170755.2 NP_001164226.1 5 569
GenBank mRNA AK097180.1 - 5 488

View Full Product Details