Gene Transcripts

Gene Symbol
Vom1r15
Entrez Gene ID
494305
Gene Name
vomeronasal 1 receptor 15
Gene Aliases
V1re22
Chromosome Location
Chr. 1: 60633626 - 60634546 on Build Rnor_6.0
UniGene ID
Rn.129452
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008960.1 NP_001008960.1 1 868
GenBank mRNA AY510361.1 - 1 868

View Full Product Details